2-Morpholinoaniline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(4-Morpholinyl)aniline
L05660-06
L05660-14
CAAAAL05660-06EA
127.19
CAD
CAAAAL05660-06
CAAAAL05660-14
2-Morpholinoaniline 98%
2-Morpholinoaniline
Purity: 98%
CAS No.: 5585-33-1
Molecular Formula: C10H14N2O
Formula Weight: 178.24
Melting Point: 95-98°
Beilstein Reference: 163926
UN No.: 2811
MDL No.: MFCD00047408
Formula:
C₁₀H₁₄N₂O Melting Pt: 95…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047408 CAS Number: 5585-33-1 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...