2-(Methylthio)ethanol 99%
Supplier: Thermo Scientific Chemicals
Warning
|
L05442-06
L05442-14
L05442-22
CAAAAL05442-06EA
46.29
CAD
CAAAAL05442-06
CAAAAL05442-14
CAAAAL05442-22
2-(Methylthio)ethanol 99%
2-(Methylthio)ethanol
Purity: 99%
CAS No.: 5271-38-5
Molecular Formula: C3H8OS
Formula Weight: 92.16
Flash Point: 70°(158°F)
Refractive Index: 1.4944
Odor: Stench
Boiling Point: 169-171°
Beilstein Reference: 1731081
UN No.: 3334
MDL No.: MFCD00002908
Formula:
C₃H₈OS Boiling Pt: 169…171 °C Density: 1.060 Flash Pt: 70 °C(158 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002908 CAS Number: 5271-38-5 EINECS: 226-090-3 UN: 3334 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...