Potassium thioacetate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Potassium ethanethioate
L05405-14
L05405-22
CAAAAL05405-14EA
95.43
CAD
CAAAAL05405-14
CAAAAL05405-22
Potassium thioacetate 98%
Potassium thioacetate
Fieser: 1,955
Purity: 98%
CAS No.: 10387-40-3
Molecular Formula: C2H3KOS
Formula Weight: 114.21
Odor: Stench
Melting Point: ca 240° dec.
Beilstein Reference: 3595448
UN No.: 3335
MDL No.: MFCD00083065
Formula:
C₂H₃KOS Melting Pt: ca 240 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00137704 CAS Number: 10387-40-3 EINECS: 233-848-7 UN: 3335 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...