7-Phenylenanthic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

L05009-03 L05009-06
CAAAAL05009-03EA 154.52 CAD
CAAAAL05009-03 CAAAAL05009-06
7-Phenylenanthic acid 97%
7-Phenylenanthic acid

Purity: 97% CAS No.: 40228-90-8 Molecular Formula: C6H5(CH2)6COOH Formula Weight: 206.29 Flash Point: 110°(230°F) Refractive Index: 1.5070 Melting Point: ca 20° Boiling Point: 147-150°/0.4mm Beilstein Reference: 1957081 MDL No.: MFCD00037801

Formula: C₁₃H₁₈O₂
Boiling Pt: 147…150 °C/0.4 mm
Melting Pt: ca 20 °C
Density: 1.009
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00037801
CAS Number: 40228-90-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR