2-Mercapto-2-thiazoline 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L04924-22
L04924-36
CAAAAL04924-22EA
62.73
CAD
CAAAAL04924-22
CAAAAL04924-36
2-Mercapto-2-thiazoline 98+%
2-Mercapto-2-thiazoline
Fieser: 11,518
Purity: 98+%
CAS No.: 96-53-7
Molecular Formula: C3H5NS2
Formula Weight: 119.21
Melting Point: 103-107°
Beilstein Reference: 106332
UN No.: 2811
MDL No.: MFCD00126013
Formula:
C₃H₅NS₂ Melting Pt: 103…107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00126013 CAS Number: 96-53-7 EINECS: 202-512-1 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...