(-)-Shikimic acid 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (3R,4S,5R)-3,4,5-Trihydroxycyclohex-1-ene-1-carboxylic acid, Shikimic acid

L04848-03 L04848-MD
CAAAAL04848-03EA 176.96 CAD
CAAAAL04848-03 CAAAAL04848-MD
(-)-Shikimic acid 98%
(-)-Shikimic acid

Antagonistic antimicrobial agent which also inhibits rapamycin biosynthesis


Purity: 98% CAS No.: 138-59-0 Molecular Formula: C7H10O5 Formula Weight: 174.16 Odor: Odorless Melting Point: 184-188° Beilstein Reference: 4782717 Merck Reference: 14,8480 Optical Rotation: -175° (c=2 in water) MDL No.: MFCD00066278

Formula: C₇H₁₀O₅
MW: 174.15 g/mol
Melting Pt: 184…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00066278
CAS Number: 138-59-0
EINECS: 205-334-2
Merck Index: 14,8480

Order Now

Specification Test Results

Optical rotation -175° (c=2 in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR