Cholesteryl chloroformate

Supplier: Thermo Scientific Chemicals
Danger

L04801-14 L04801-22
CAAAAL04801-14EA 81.53 CAD
CAAAAL04801-14 CAAAAL04801-22
Cholesteryl chloroformate
Cholesteryl chloroformate

CAS No.: 7144-08-3 Molecular Formula: C28H45ClO2 Formula Weight: 449.12 Storage Temperature: 2°C to 8°C Odor: Pungent Melting Point: 116-120° Beilstein Reference: 3223154 Optical Rotation: -29° (c=5 in chloroform) UN No.: 3261 MDL No.: MFCD00003633

Formula: C₂₈H₄₅ClO₂
Melting Pt: 116…120 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003633
CAS Number: 7144-08-3
EINECS: 230-447-9
UN: 3261
ADR: "8",III

Order Now

Specification Test Results

Optical rotation -29° (c=5 in chloroform)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR