Dimethylphenylsilane 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dimethyl(phenyl)silane

L04558-09 L04558-18
CAAAAL04558-09EA 89.27 CAD
CAAAAL04558-09 CAAAAL04558-18
Dimethylphenylsilane 97%
Dimethylphenylsilane

Fieser: 12,209 13,123 15,168 21,218


Purity: 97% CAS No.: 766-77-8 Molecular Formula: C8H12Si Formula Weight: 136.27 Flash Point: 35°(95°F) Refractive Index: 1.4970 Melting Point: -124° Boiling Point: 156-157° Beilstein Reference: 2204906 UN No.: 1993 MDL No.: MFCD00008257

Formula: C₈H₁₂Si
MW: 136.27 g/mol
Boiling Pt: 156…157 °C
Melting Pt: –124 °C
Density: 0.879
Flash Pt: 35 °C (95 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008257
CAS Number: 766-77-8
EINECS: 212-170-5
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR