2-Iodoacetyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L04428-06
L04428-14
CAAAAL04428-06EA
216.46
CAD
CAAAAL04428-06
CAAAAL04428-14
2-Iodoacetyl chloride 97%
2-Iodoacetyl chloride
Purity: 97%
CAS No.: 38020-81-4
Molecular Formula: ICH2COCl
Formula Weight: 204.39
Storage Temperature: 2°C to 8°C
UN No.: 3265
MDL No.: MFCD00013718
Formula:
C₂H₂ClIO Boiling Pt: 48…50 °C/12 mm Density: 2.280 Storage Temperature: Refrigerator |
MDL Number:
MFCD00013718 CAS Number: 38020-81-4 EINECS: 253-748-7 UN: 3265 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...