(Ethylthio)acetone 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L04142-06
L04142-14
CAAAAL04142-06EA
79.47
CAD
CAAAAL04142-06
CAAAAL04142-14
(Ethylthio)acetone 95%
(Ethylthio)acetone
Purity: 95%
CAS No.: 20996-62-7
Molecular Formula: C5H10OS
Formula Weight: 118.20
Flash Point: 52°(125°F)
Refractive Index: 1.4720
Boiling Point: 170-172°
Beilstein Reference: 1740830
UN No.: 1993
MDL No.: MFCD00026976
Formula:
C₅H₁₀OS Boiling Pt: 170…172 °C Flash Pt: 52 °C (125 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00026976 CAS Number: 20996-62-7 EINECS: 244-143-9 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...