Triethylene glycol dimethyl ether 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,5,8,11-Tetraoxadodecane

L04128-0E L04128-22 L04128-36
CAAAAL04128-0EEA 395 CAD
CAAAAL04128-0E CAAAAL04128-22 CAAAAL04128-36
Triethylene glycol dimethyl ether 99%
Triethylene glycol dimethyl ether

Purity: 99% CAS No.: 112-49-2 Molecular Formula: C8H18O4 Formula Weight: 178.23 Flash Point: 113°(235°F) Refractive Index: 1.4230 Melting Point: -45° Boiling Point: 215-216° Beilstein Reference: 1700630 Merck Reference: 14,9692 MDL No.: MFCD00008504

Formula: C₈H₁₈O₄
MW: 178.23 g/mol
Boiling Pt: 215…216 °C
Melting Pt: –45 °C
Density: 0.986
Flash Pt: 113 °C(235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008504
CAS Number: 112-49-2
EINECS: 203-977-3
Merck Index: 14,9692

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR