N,N-Dimethyl-1,3-diaminopropane 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-Dimethylaminopropylamine, N,N-Dimethyltrimethylenediamine, N,N-Dimethyl-1,3-propanediamine
L03539-AK
L03539-AU
CAAAAL03539-AKEA
40.92
CAD
CAAAAL03539-AK
CAAAAL03539-AU
N,N-Dimethyl-1,3-diaminopropane 99%
N,N-Dimethyl-1,3-diaminopropane
Fieser: 1,274
Purity: 99%
CAS No.: 109-55-7
Molecular Formula: C5H14N2
Formula Weight: 102.18
Flash Point: 32°(89°F)
Refractive Index: 1.4350
Odor: Ammonia-like
Melting Point: -70°
Boiling Point: 126-128°
Beilstein Reference: 605293
UN No.: 2734
MDL No.: MFCD00008216
Formula:
C₅H₁₄N₂ MW: 102.18 g/mol Boiling Pt: 126…128 °C Melting Pt: –70 °C Density: 0.818 Flash Pt: 32 °C (89 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008216 CAS Number: 109-55-7 EINECS: 203-680-9 UN: 2734 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...