N,N-Dimethylcarbamoyl chloride 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L03415-14
L03415-22
CAAAAL03415-14EA
57.04
CAD
CAAAAL03415-14
CAAAAL03415-22
N,N-Dimethylcarbamoyl chloride 96%
N,N-Dimethylcarbamoyl chloride
Special handling precautions required. View MSDS prior to purchase.
Purity: 96%
CAS No.: 79-44-7
Molecular Formula: C3H6ClNO
Formula Weight: 107.54
Flash Point: 68°(154°F)
Refractive Index: 1.4530
Odor: Unpleasant
Melting Point: -33°
Boiling Point: 167-168°
Beilstein Reference: 878197
UN No.: 2262
MDL No.: MFCD00000635
Caution: Special handling precautions required. View MSDS prior to purchase.
Formula:
C₃H₆ClNO Boiling Pt: 167…168 °C Melting Pt: –33 °C Density: 1.171 Flash Pt: 68 °C(154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000635 CAS Number: 79-44-7 EINECS: 201-208-6 UN: 2622 ADR: "3",II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...