(Methylthio)acetonitrile 99%

Supplier: Thermo Scientific Chemicals
Warning

L03066-06 L03066-14 L03066-22
CAAAAL03066-06EA 46.45 CAD
CAAAAL03066-06 CAAAAL03066-14 CAAAAL03066-22
(Methylthio)acetonitrile 99%
(Methylthio)acetonitrile

Fieser: 8,349


Purity: 99% CAS No.: 35120-10-6 Molecular Formula: C3H5NS Formula Weight: 87.14 Flash Point: 67°(152°F) Refractive Index: 1.4820 Boiling Point: 166-167° Beilstein Reference: 1738015 MDL No.: MFCD00001925

Formula: C₃H₅NS
Boiling Pt: 166…167 °C
Density: 1.052
Flash Pt: 67 °C(152 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001925
CAS Number: 35120-10-6
EINECS: 252-383-0
UN: 2810
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR