Diuron (N-(3,4-dichlorophenyl)-N',N'-dimethylurea) 97%

Supplier: Thermo Scientific Chemicals
Warning

L02986-18 L02986-30
CAAAAL02986-18EA 72.05 CAD
CAAAAL02986-18 CAAAAL02986-30
Diuron (N-(3,4-dichlorophenyl)-N',N'-dimethylurea) 97%
Diuron (N-(3,4-Dichlorophenyl)-N',N'-dimethylurea)

Purity: 97% CAS No.: 330-54-1 Molecular Formula: C9H10Cl2N2O Formula Weight: 233.10 Odor: Odorless Melting Point: 157-160° Boiling Point: 180-190° dec. Beilstein Reference: 2215168 Merck Reference: 14,3382 UN No.: 3077 MDL No.: MFCD00018136

Formula: C₉H₁₀Cl₂N₂O
MW: 233.10 g/mol
Boiling Pt: 180…190 °C (dec.)
Melting Pt: 157…160 °C
Density: 1.48
Storage Temperature: Ambient
MDL Number: MFCD00018136
CAS Number: 330-54-1
EINECS: 206-354-4
UN: 3077
ADR: "9",III
Merck Index: 14,3382

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR