1-Butyl-2-urea 96%

Supplier: Thermo Scientific Chemicals
Warning

L02854-22 L02854-36
CAAAAL02854-22EA 316 CAD
CAAAAL02854-22 CAAAAL02854-36
1-Butyl-2-urea 96%
1-Butyl-2-urea

Purity: 96% CAS No.: 592-31-4 Molecular Formula: CH3(CH2)3NHCONH2 Formula Weight: 116.16 Melting Point: 92-96° Beilstein Reference: 1744775

Formula: C₅H₁₂N₂O
Melting Pt: 92…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00007956
CAS Number: 592-31-4
EINECS: 209-748-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR