3,4-Dihydro-2H-pyran 99%

Supplier: Thermo Scientific Chemicals
Danger

L02731-AC L02731-AE L02731-AP
CAAAAL02731-ACEA 45.66 CAD
CAAAAL02731-AC CAAAAL02731-AE CAAAAL02731-AP
3,4-Dihydro-2H-pyran 99%
3,4-Dihydro-2H-pyran

Widely used hydroxyl-protecting reagent


Purity: 99% CAS No.: 110-87-2 Molecular Formula: C5H8O Formula Weight: 84.12 Flash Point: -15°(5°F) Refractive Index: 1.4420 Odor: Unpleasant pH: 7.0 Melting Point: -70° Boiling Point: 85-86° Beilstein Reference: 103493 UN No.: 2376 MDL No.: MFCD00006558

Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 85…86 °C
Melting Pt: –70 °C
Density: 0.926
Flash Pt: -15 °C(5 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006558
CAS Number: 110-87-2
EINECS: 203-810-4
UN: 2376
ADR: "3",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR