(Methylamino)acetaldehyde dimethylacetal 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,2-Dimethoxy-N-methylethanamine
L02391-14
L02391-22
CAAAAL02391-14EA
73
CAD
CAAAAL02391-14
CAAAAL02391-22
(Methylamino)acetaldehyde dimethylacetal 97%
(Methylamino)acetaldehyde dimethylacetal
Purity: 97%
CAS No.: 122-07-6
Molecular Formula: CH3NHCH2CH(OCH3)2
Formula Weight: 119.16
Flash Point: 29°(84°F)
Refractive Index: 1.4115
Odor: Ammonia-like
pH: 10.9
Melting Point: -73°
Boiling Point: 139-140°
Beilstein Reference: 605322
UN No.: 1993
MDL No.: MFCD00008485
Formula:
C₅H₁₃NO₂ Boiling Pt: 139…140 °C Melting Pt: –73 °C Density: 0.928 Flash Pt: 29 °C(84 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008485 CAS Number: 122-07-6 EINECS: 204-520-0 UN: 3271 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...