Tropine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: endo-8-Methyl-8-azabicyclo[3.2.1]octan-3-ol

L02332-09 L02332-18
CAAAAL02332-09EA 74.26 CAD
CAAAAL02332-09 CAAAAL02332-18
Tropine 98%
Tropine

Purity: 98% CAS No.: 120-29-6 Molecular Formula: C8H15NO Formula Weight: 141.22 Storage Temperature: 2°C to 8°C Odor: Amine-like pH: 12 Melting Point: 61-65° (dried) Boiling Point: 232-234° Beilstein Reference: 80188 Merck Reference: 14,9781 MDL No.: MFCD00005551

Formula: C₈H₁₅NO
MW: 141.21 g/mol
Boiling Pt: 232…234 °C
Melting Pt: 61…65 °C (dried)
Density: 1.04
Storage Temperature: Refrigerator
MDL Number: MFCD00180667
CAS Number: 120-29-6
EINECS: 204-384-2
Merck Index: 14,9781

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR