Hexaethylene glycol 96%

Supplier: Thermo Scientific Chemicals

Synonyms: 3,6,9,12,15-Pentaoxaheptadecane-1,17-diol

L02161-06 L02161-14
CAAAAL02161-06EA 116.6 CAD
CAAAAL02161-06 CAAAAL02161-14
Hexaethylene glycol 96%
Hexaethylene glycol

Purity: 96% CAS No.: 2615-15-8 Molecular Formula: H(OCH2CH2)6OH Formula Weight: 282.33 Flash Point: 110°(230°F) Refractive Index: 1.4640 Melting Point: 5-7° Boiling Point: 216-217°/4mm Beilstein Reference: 1638281 MDL No.: MFCD00002877

Formula: C₁₂H₂₆O₇
Boiling Pt: 216…217 °C/4 mm
Melting Pt: 5…7 °C
Density: 1.123
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002877
CAS Number: 2615-15-8
EINECS: 220-045-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR