1,2-Bis(phenylthio)ethane 98+%
Supplier: Thermo Scientific Chemicals
L02019-09
L02019-18
CAAAAL02019-09EA
96.85
CAD
CAAAAL02019-09
CAAAAL02019-18
1,2-Bis(phenylthio)ethane 98+%
1,2-Bis(phenylthio)ethane
Purity: 98+%
CAS No.: 622-20-8
Molecular Formula: C14H14S2
Formula Weight: 246.39
Melting Point: 68-70°
Beilstein Reference: 1877719
Formula:
C₁₄H₁₄S₂ Melting Pt: 68…70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014085 CAS Number: 622-20-8 EINECS: 210-723-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...