Tropinone 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 8-Methyl-8-azabicyclo[3.2.1]octan-3-one

L02011-06 L02011-14 L02011-22
CAAAAL02011-06EA 56.72 CAD
CAAAAL02011-06 CAAAAL02011-14 CAAAAL02011-22
Tropinone 99%
Tropinone

Purity: 99% CAS No.: 532-24-1 Molecular Formula: C8H13NO Formula Weight: 139.20 Storage Temperature: 2°C to 8°C Flash Point: 90°(194°F) Odor: Odorless Melting Point: 40-43° Boiling Point: 227° Beilstein Reference: 2329 MDL No.: MFCD00005549

Formula: C₈H₁₃NO
MW: 139.2 g/mol
Boiling Pt: 227 °C
Melting Pt: 40…43 °C
Flash Pt: 90 °C(194 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00005549
CAS Number: 532-24-1
EINECS: 208-530-6
UN: 3259
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR