α-Fluorophenylacetic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

L01745-04 L01745-MC L01745-ME
CAAAAL01745-04EA 303.36 CAD
CAAAAL01745-04 CAAAAL01745-MC CAAAAL01745-ME
α-Fluorophenylacetic acid 98%
α-Fluorophenylacetic acid

Purity: 98% CAS No.: 1578-63-8 Molecular Formula: C6H5CHFCO2H Formula Weight: 154.14 Melting Point: 78-82° Beilstein Reference: 2087378 UN No.: 2811 MDL No.: MFCD00004221

Formula: C₈H₇FO₂
Melting Pt: 78…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00004221
CAS Number: 1578-63-8
EINECS: 216-420-4
UN: 2811
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR