Bis(phenylthio)methane 98+%

Supplier: Thermo Scientific Chemicals

L01723-06 L01723-14
CAAAAL01723-06EA 52.61 CAD
CAAAAL01723-06 CAAAAL01723-14
Bis(phenylthio)methane 98+%
Bis(phenylthio)methane

Fieser: 6,267 7,25 10,42 14,50


Purity: 98+% CAS No.: 3561-67-9 Molecular Formula: C13H12S2 Formula Weight: 232.36 Melting Point: 36-38° Boiling Point: 194°/8mm MDL No.: MFCD00003067

Formula: C₁₃H₁₂S₂
MW: 232.37 g/mol
Boiling Pt: 194 °C/8 mm
Melting Pt: 36…38 °C
Storage Temperature: Ambient
MDL Number: MFCD00003067
CAS Number: 3561-67-9
EINECS: 222-624-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR