N,N-Dimethylacetamide dimethyl acetal tech. 90% stabilized, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stabilization:
Stabilised with methanol 5-10%
L01642-06
L01642-14
CAAAAL01642-06EA
87.37
CAD
CAAAAL01642-06
CAAAAL01642-14
N,N-Dimethylacetamide dimethyl acetal tech. 90% stabilized, Technical Grade
N,N-Dimethylacetamide dimethyl acetal
Fieser: 4,166 5,226 10,154
Purity: 90%
CAS No.: 18871-66-4
Molecular Formula: C6H15NO2
Formula Weight: 133.19
Flash Point: 8°(46°F)
Refractive Index: 1.4120
Boiling Point: 118-120°
Beilstein Reference: 1420865
UN No.: 1993
MDL No.: MFCD00008476
Formula:
C₆H₁₅NO₂ Boiling Pt: 118…120 °C Density: 0.911 Flash Pt: 8 °C(46 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008476 CAS Number: 18871-66-4 EINECS: 242-641-0 UN: 1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...