Valeric acid hydrazide 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pentanehydrazide

L01555-06 L01555-14
CAAAAL01555-06EA 44.08 CAD
CAAAAL01555-06 CAAAAL01555-14 CAAAAL01555-22
Valeric acid hydrazide 95%
Valeric acid hydrazide

Purity: 95% CAS No.: 38291-82-6 Molecular Formula: CH3(CH2)3CONHNH2 Formula Weight: 116.16 Melting Point: 55-64° Beilstein Reference: 1744747

Formula: C₅H₁₂N₂O
MW: 116.16 g/mol
Melting Pt: 55…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00025137
CAS Number: 38291-82-6
EINECS: 253-864-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR