1,3-Bis(trimethylsilyl)urea 98+%
Supplier: Thermo Scientific Chemicals
Synonyms:
N,N'-Bis(trimethylsilyl)urea
L01301-18
L01301-30
CAAAAL01301-18EA
43.61
CAD
CAAAAL01301-18
CAAAAL01301-30
1,3-Bis(trimethylsilyl)urea 98+%
1,3-Bis(trimethylsilyl)urea
Fieser: 11,68
Purity: 98+%
CAS No.: 18297-63-7
Molecular Formula: C7H20N2OSi2
Formula Weight: 204.42
Melting Point: ca 230° dec.
Beilstein Reference: 1937452
Formula:
C₇H₂₀N₂OSi₂ MW: 204.42 g/mol Melting Pt: ∼ 230 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00008260 CAS Number: 18297-63-7 EINECS: 242-177-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...