3-Chlorobenzo[b]thiophene-2-carboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L01257-03
L01257-06
CAAAAL01257-03EA
60.83
CAD
CAAAAL01257-03
CAAAAL01257-06
3-Chlorobenzo[b]thiophene-2-carboxylic acid 97%
3-Chlorobenzo[b]thiophene-2-carboxylic acid
Purity: 97%
CAS No.: 21211-22-3
Molecular Formula: C9H5ClO2S
Formula Weight: 212.65
Melting Point: 265-267°
Beilstein Reference: 1425602
Formula:
C₉H₅ClO₂S Melting Pt: 265…267 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014579 CAS Number: 21211-22-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...