2-Amino-1,3,4-thiadiazole 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L01244-06
L01244-14
CAAAAL01244-06EA
67.31
CAD
CAAAAL01244-06
CAAAAL01244-14
2-Amino-1,3,4-thiadiazole 98+%
2-Amino-1,3,4-thiadiazole
Purity: 98+%
CAS No.: 4005-51-0
Molecular Formula: C2H3N3S
Formula Weight: 101.13
Melting Point: 188-194°
Beilstein Reference: 107135
UN No.: 2811
MDL No.: MFCD00003107
Formula:
C₂H₃N₃S Melting Pt: 188…194 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003107 CAS Number: 4005-51-0 EINECS: 223-657-7 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...