(1-Naphthoxy)acetic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(Naphthalen-1-yloxy)acetic acid
L01036-06
L01036-14
CAAAAL01036-06EA
67.62
CAD
CAAAAL01036-06
CAAAAL01036-14
(1-Naphthoxy)acetic acid 98+%
(1-Naphthoxy)acetic acid
Purity: 98+%
CAS No.: 2976-75-2
Molecular Formula: C12H10O3
Formula Weight: 202.21
Melting Point: 193-197°
Beilstein Reference: 1912165
Formula:
C₁₂H₁₀O₃ MW: 202.21 g/mol Melting Pt: 193…197 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003927 CAS Number: 2976-75-2 EINECS: 221-023-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...