Phenyl carbamate 98+%
Supplier: Thermo Scientific Chemicals
L00944-06
L00944-14
CAAAAL00944-06EA
72.68
CAD
CAAAAL00944-06
CAAAAL00944-14
Phenyl carbamate 98+%
Phenyl carbamate
Purity: 98+%
CAS No.: 622-46-8
Molecular Formula: H2NCO2C6H5
Formula Weight: 137.14
Melting Point: 149-152°
Beilstein Reference: 2042627
Formula:
C₇H₇NO₂ Melting Pt: 149…152 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007961 CAS Number: 622-46-8 EINECS: 210-737-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...