O-Phenyl chlorothioformate 98+%

Supplier: Thermo Scientific Chemicals
Danger

L00838-03 L00838-06 L00838-14
CAAAAL00838-03EA 49.93 CAD
CAAAAL00838-03 CAAAAL00838-06 CAAAAL00838-14
O-Phenyl chlorothioformate 98+%
O-Phenyl chlorothioformate

Fieser: 10,306 21,341


Purity: 98+% CAS No.: 1005-56-7 Molecular Formula: C7H5ClOS Formula Weight: 172.63 Storage Temperature: 2°C to 8°C Flash Point: 81°(177°F) Refractive Index: 1.5804 Odor: Acrid Boiling Point: 81-83°/6mm Beilstein Reference: 774830 UN No.: 3265 MDL No.: MFCD00004920

Formula: C₇H₅ClOS
Boiling Pt: 81…83 °C/6 mm
Density: 1.248
Flash Pt: 81 °C(177 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00004920
CAS Number: 1005-56-7
EINECS: 213-736-4
UN: 3265
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR