Triphenylvinylphosphonium bromide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L00708-06
L00708-14
L00708-22
CAAAAL00708-06EA
110.6
CAD
CAAAAL00708-06
CAAAAL00708-14
CAAAAL00708-22
Triphenylvinylphosphonium bromide 97%
Triphenylvinylphosphonium bromide
Fieser: 1,1274 2,456 5,750 6,666 11,596 13,343 19,398
Purity: 97%
CAS No.: 5044-52-0
Molecular Formula: CH2=CHP(C5H5)3Br
Formula Weight: 369.25
Melting Point: 182-186°
Beilstein Reference: 4168860
Formula:
C₂₀H₁₈BrP Melting Pt: 182…186 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011807 CAS Number: 5044-52-0 EINECS: 225-740-3 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...