Chlorodimethyl(pentafluorophenyl)silane (flophemesyl chloride) 96%

Supplier: Thermo Scientific Chemicals
Danger

L00550-03 L00550-06
CAAAAL00550-03EA 66.83 CAD
CAAAAL00550-03 CAAAAL00550-06
Chlorodimethyl(pentafluorophenyl)silane (flophemesyl chloride) 96%
Chlorodimethyl(pentafluorophenyl)silane (Flophemesyl chloride)

Purity: 96% CAS No.: 20082-71-7 Molecular Formula: C8H6ClF5Si Formula Weight: 260.66 Flash Point: 95°(203°F) Refractive Index: 1.4470 Boiling Point: 78-81°/10mm Beilstein Reference: 2858407 UN No.: 2987 MDL No.: MFCD00000500

Formula: C₈H₆ClF₅Si
Boiling Pt: 78…81 °C/10 mm
Density: 1.381
Flash Pt: 95 °C(203 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000500
CAS Number: 20082-71-7
EINECS: 243-506-9
UN: 2987
ADR: "8",II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR