2,2-Dimethyl-1,3-dioxolane 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L00482-06
L00482-14
CAAAAL00482-06EA
52.3
CAD
CAAAAL00482-06
CAAAAL00482-14
2,2-Dimethyl-1,3-dioxolane 98+%
2,2-Dimethyl-1,3-dioxolane
Purity: 98+%
CAS No.: 2916-31-6
Molecular Formula: C5H10O2
Formula Weight: 102.13
Flash Point: 6°(42°F)
Refractive Index: 1.3980
Boiling Point: 91-93°
Beilstein Reference: 102775
UN No.: 1993
MDL No.: MFCD00014106
Formula:
C₅H₁₀O₂ MW: 102.13 g/mol Boiling Pt: 91…93 °C Density: 0.946 Flash Pt: 6 °C(42 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014106 CAS Number: 2916-31-6 EINECS: 220-842-4 UN: 1993 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...