2,4,6-Trimethylpyrylium tetrafluoroborate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L00467-06
L00467-14
CAAAAL00467-06EA
108.23
CAD
CAAAAL00467-06
CAAAAL00467-14
2,4,6-Trimethylpyrylium tetrafluoroborate 98+%
2,4,6-Trimethylpyrylium tetrafluoroborate
Purity: 98+%
CAS No.: 773-01-3
Molecular Formula: C8H11BF4O
Formula Weight: 209.98
Storage Temperature: 2°C to 8°C
Melting Point: 224-226°
Beilstein Reference: 3639361
MDL No.: MFCD00032098
Formula:
C₈H₁₁BF₄O Melting Pt: 224…226 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00032098 CAS Number: 773-01-3 EINECS: 212-256-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...