Di-tert-butyl azodicarboxylate 98%

Supplier: Thermo Scientific Chemicals
Warning

L00294-06 L00294-14
CAAAAL00294-06EA 79.47 CAD
CAAAAL00294-06 CAAAAL00294-14
Di-tert-butyl azodicarboxylate 98%
Di-tert-butyl azodicarboxylate

Fieser: 1,209 14,115 18,119


Purity: 98% CAS No.: 870-50-8 Molecular Formula: (CH3)3CO2C-N=N-CO2C(CH3)3 Formula Weight: 230.27 Melting Point: 89-92° Beilstein Reference: 1911434

Formula: C₁₀H₁₈N₂O₄
Melting Pt: 89…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00015001
CAS Number: 870-50-8
EINECS: 212-796-9
UN: 1325
ADR: "4.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR