Diphenylacetyl chloride 90+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L00171-09
L00171-18
CAAAAL00171-09EA
39.03
CAD
CAAAAL00171-09
CAAAAL00171-18
Diphenylacetyl chloride 90+%
Diphenylacetyl chloride
Fieser: 20,157
Purity: 90+%
CAS No.: 1871-76-7
Molecular Formula: C14H11ClO
Formula Weight: 230.69
Melting Point: 50-56°
Boiling Point: 175-176°/17mm
Merck Reference: 14,3316
UN No.: 3261
MDL No.: MFCD00013655
Formula:
C₁₄H₁₁ClO Boiling Pt: 175…176 °C/17 mm Melting Pt: 50…56 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013655 CAS Number: 1871-76-7 EINECS: 217-493-5 UN: 3261 ADR: 8,III Merck Index: 14,3316 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...