N-(Hydroxymethyl)-2,2,2-trifluoroacetamide 98+%

Supplier: Thermo Scientific Chemicals
Warning

L00128-06 L00128-14
CAAAAL00128-06EA 102.7 CAD
CAAAAL00128-06 CAAAAL00128-14
N-(Hydroxymethyl)-2,2,2-trifluoroacetamide 98+%
N-(Hydroxymethyl)-2,2,2-trifluoroacetamide

Purity: 98+% CAS No.: 50667-69-1 Molecular Formula: C3H4F3NO2 Formula Weight: 143.06 Storage Temperature: 2°C to 8°C Melting Point: 99-103° Beilstein Reference: 1859914 MDL No.: MFCD00014416

Formula: C₃H₄F₃NO₂
Melting Pt: 99…103 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00014416
CAS Number: 50667-69-1
EINECS: 256-698-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR