2-Propyl-1-heptanol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H50340-03
H50340-06
CAAAAH50340-03EA
333.38
CAD
CAAAAH50340-03
CAAAAH50340-06
2-Propyl-1-heptanol 98%
2-Propyl-1-heptanol
Purity: 98%
CAS No.: 10042-59-8
Molecular Formula: C10H22O
Formula Weight: 158.28
MDL No.: MFCD00046768
Formula:
C₁₀H₂₂O Storage Temperature: Ambient |
MDL Number:
MFCD00046768 CAS Number: 10042-59-8 EINECS: 233-126-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...