Glycine propyl ester hydrochloride

Supplier: Thermo Scientific Chemicals
Warning

H50250-03 H50250-06
CAAAAH50250-03EA 63.83 CAD
CAAAAH50250-03 CAAAAH50250-06
Glycine propyl ester hydrochloride
Glycine propyl ester hydrochloride

CAS No.: 13049-01-9 Molecular Formula: H2NCH2CO2(CH2)2CH3.HCl Formula Weight: 117.15 Melting Point: 78-80° MDL No.: MFCD00151829

Formula: C₅H₁₂ClNO₂
Melting Pt: 78…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00151829
CAS Number: 13049-01-9
EINECS: 235-925-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR