Glycine iso-propyl ester hydrochloride (H-Gly-OiPr.HCl) 96%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Gly-OIpr·HCl, Isopropyl 2-aminoacetate hydrochloride

H50249-03 H50249-06
CAAAAH50249-03EA 69.68 CAD
CAAAAH50249-03 CAAAAH50249-06
Glycine iso-propyl ester hydrochloride (H-Gly-OiPr.HCl) 96%
Glycine iso-propyl ester hydrochloride (H-Gly-OiPr.HCl)

Purity: 96% CAS No.: 14019-62-6 Molecular Formula: C5H11NO2.HCl Formula Weight: 153.61 MDL No.: MFCD00151826

Formula: C₅H₁₂ClNO₂
MW: 153.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151826
CAS Number: 14019-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR