Glycine benzyl ester hydrochloride (H-Gly-OBzl.HCl) 98%
Supplier: Thermo Scientific Chemicals
H50247-03
H50247-06
CAAAAH50247-03EA
55.93
CAD
CAAAAH50247-03
CAAAAH50247-06
Glycine benzyl ester hydrochloride (H-Gly-OBzl.HCl) 98%
Glycine benzyl ester hydrochloride (H-Gly-OBzl.HCl)
Purity: 98%
CAS No.: 2462-31-9
Molecular Formula: C9H11NO2.HCl
Formula Weight: 201.65
Storage Temperature: 2°C to 8°C
Melting Point: 139-140°
MDL No.: MFCD00035442
Formula:
C₉H₁₂ClNO₂ Melting Pt: 139…140 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00035442 CAS Number: 2462-31-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...