1-Chlorophthalazine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H27143-03
H27143-09
CAAAH27143-03EA
93.22
CAD
CAAAH27143-03
CAAAH27143-09
1-Chlorophthalazine 97%
1-Chlorophthalazine
Purity: 97%
CAS No.: 5784-45-2
Molecular Formula: C8H5N2Cl
Formula Weight: 164.59
Storage Temperature: 2°C to 8°C
Melting Point: 109-112°
MDL No.: MFCD00024141
Formula:
C₈H₅ClN₂ Melting Pt: 109…112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00024141 CAS Number: 5784-45-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...