(±)-1,2,3,4-Tetrahydro-1-naphthylamine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tetralin-1-amine, 1,2,3,4-Tetrahydronaphthalen-1-amine

B25673-06 B25673-14
CAAAAB25673-06EA 77.89 CAD
CAAAAB25673-06 CAAAAB25673-14
(±)-1,2,3,4-Tetrahydro-1-naphthylamine 97%
(±)-1,2,3,4-Tetrahydro-1-naphthylamine

Purity: 97% CAS No.: 2217-40-5 Molecular Formula: C10H13N Formula Weight: 147.22 Flash Point: 110°(230°F) Refractive Index: 1.5640 Boiling Point: 246-247°/714mm MDL No.: MFCD00001740

Formula: C₁₀H₁₃N
Boiling Pt: 246…247 °C/714 mm
Density: 1.026
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001740
CAS Number: 2217-40-5
EINECS: 218-712-7
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR