1-(1-Cyclohexen-1-yl)pyrrolidine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25622-06
B25622-14
B25622-22
CAAAAB25622-06EA
60.83
CAD
CAAAAB25622-06
CAAAAB25622-14
CAAAAB25622-22
1-(1-Cyclohexen-1-yl)pyrrolidine 97%
1-(1-Cyclohexen-1-yl)pyrrolidine
Purity: 97%
CAS No.: 1125-99-1
Molecular Formula: C10H17N
Formula Weight: 151.25
Storage Temperature: 2°C to 8°C
Flash Point: 66°(151°F)
Refractive Index: 1.5217
Boiling Point: 114-115°/15mm
Beilstein Reference: 114797
MDL No.: MFCD00003163
Formula:
C₁₀H₁₇N Boiling Pt: 114…115 °C (15 mmHg) Density: 0,940 Flash Pt: 66 °C (151 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00003163 CAS Number: 1125-99-1 EINECS: 214-414-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...