(±)-Indoline-2-carboxylic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Indoline-2-carboxylic acid

B25450-06 B25450-14
CAAAAB25450-06EA 98.43 CAD
CAAAAB25450-06 CAAAAB25450-14
(±)-Indoline-2-carboxylic acid 95%
(±)-Indoline-2-carboxylic acid

Purity: 95% CAS No.: 78348-24-0 Molecular Formula: C9H9NO2 Formula Weight: 163.18 Melting Point: ca 168° dec. MDL No.: MFCD00010635

Formula: C₉H₉NO₂
MW: 163.18 g/mol
Melting Pt: ∼ 168 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00010635
CAS Number: 78348-24-0
EINECS: 278-899-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR