Dibenzyl succinate 98%
Supplier: Thermo Scientific Chemicals
B25257-14
B25257-22
CAAAAB25257-14EA
129.72
CAD
CAAAAB25257-14
CAAAAB25257-22
CAAAAB25257-36
Dibenzyl succinate 98%
Dibenzyl succinate
Purity: 98%
CAS No.: 103-43-5
Molecular Formula: C18H18O4
Formula Weight: 298.34
Refractive Index: 1.5960
Melting Point: 42-47°
Boiling Point: 245°/15mm
Beilstein Reference: 1916056
MDL No.: MFCD00022034
Formula:
C₁₈H₁₈O₄ MW: 298.34 g/mol Boiling Pt: 245 °C (15 mmHg) Melting Pt: 42…47 °C Density: 1.256 Storage Temperature: Ambient |
MDL Number:
MFCD00022034 CAS Number: 103-43-5 EINECS: 203-110-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...