CAPSO sodium salt (3-(cyclohexylamino)-2-hydroxypropanesulfonic acid sodium salt) 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium 3-(cyclohexylamino)-2-hydroxypropane-1-sulfonate, Sodium 3-(cyclohexylamino)-2-hydroxypropane-1-sulphonate

B25202-14 B25202-22 B25202-36
CAAAAB25202-14EA 64.78 CAD
CAAAAB25202-14 CAAAAB25202-22 CAAAAB25202-36
CAPSO sodium salt (3-(cyclohexylamino)-2-hydroxypropanesulfonic acid sodium salt) 98%
CAPSO sodium salt (3-(Cyclohexylamino)-2-hydroxypropanesulphonic acid sodium salt)

Purity: 98% CAS No.: 102601-34-3 Molecular Formula: C9H18NNaO4S Formula Weight: 259.30 MDL No.: MFCD00070063

Formula: C₉H₁₈NNaO₄S
MW: 259.30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070063
CAS Number: 102601-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR