Glyoxylic acid 50% (w/w) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Oxoacetic acid

B25149-0B B25149-0I B25149-30
CAAAAB25149-0BEA 192.76 CAD
CAAAAB25149-0B CAAAAB25149-0I CAAAAB25149-30
Glyoxylic acid 50% (w/w) in aqueous solution
Glyoxylic acid

CAS No.: 298-12-4 Molecular Formula: OHCCO2H Formula Weight: 74.04 Storage Temperature: 2°C to 8°C Melting Point: -93° Boiling Point: 111° UN No.: 3265 MDL No.: MFCD00006958

Formula: C₂H₂O₃
MW: 74.04 g/mol
Boiling Pt: 111 °C
Melting Pt: –93 °C
Density: 1.339
Storage Temperature: Ambient
MDL Number: MFCD00006958
CAS Number: 298-12-4
EINECS: 206-058-5
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR